Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Parp10
Entrez Gene ID
671535
Gene Name
poly (ADP-ribose) polymerase family, member 10
Gene Aliases
-
Chromosome Location
Chr.15: 76232995 - 76243440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163575.1 NP_001157047.1 6 - 7 1651 65
NM_001163576.1 NP_001157048.1 5 - 6 1755 65
GenBank mRNA AK028373.1 - 6 - 7 1599 65
AK035309.1 - 5 - 6 1725 65
AK046755.1 - 5 - 6 1736 65
AK153205.1 - 5 - 6 1492 65
AK166291.1 - 6 - 7 1602 65
BC023056.1 - 2 - 3 488 65
BC024074.1 - 6 - 7 1608 65
BC025608.1 - 2 - 3 633 65

View Full Product Details