Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a8a
Entrez Gene ID
64381
Gene Name
membrane-spanning 4-domains, subfamily A, member 8A
Gene Aliases
2010004L09Rik, AI481240, CD20L5, Ms4a8
Chromosome Location
Chr.19: 11067471 - 11081103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022430.2 NP_071875.2 1 - 2 122 98
GenBank mRNA AF237906.1 - 1 - 2 60 98
AK008099.1 - 1 - 2 62 98
AK172700.1 - 1 - 2 123 98
AY039038.1 - 1 - 2 60 98
BC042719.1 - 1 - 2 108 98
CV305829.1 - 1 - 2 122 98

View Full Product Details