Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a6c
Entrez Gene ID
73656
Gene Name
membrane-spanning 4-domains, subfamily A, member 6C
Gene Aliases
2200009H22Rik, 2210417N07Rik
Chromosome Location
Chr.19: 11469041 - 11487069 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028595.4 NP_082871.2 4 - 5 543 86
GenBank mRNA AF237910.1 - 4 - 5 501 86
AK150397.1 - 4 - 5 440 86
AK152258.1 - 4 - 5 440 86

View Full Product Details