Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stx3
Entrez Gene ID
20908
Gene Name
syntaxin 3
Gene Aliases
Syn-3
Chromosome Location
Chr.19: 11775118 - 11820422 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025307.1 NP_001020478.1 10 - 11 1555 101
NM_001286543.1 NP_001273472.1 11 - 12 1601 101
NM_011502.3 NP_035632.1 10 - 11 1501 101
GenBank mRNA AK038415.1 - 10 - 11 1062 101
BC024844.1 - 10 - 11 963 101
BC056949.1 - 11 - 12 1601 101
D29800.1 - 11 - 12 933 101

View Full Product Details