Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbas
Entrez Gene ID
14467
Gene Name
glioblastoma amplified sequence
Gene Aliases
AV006093, Nipsnap2
Chromosome Location
Chr.5: 129725044 - 129758327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008095.4 NP_032121.3 3 - 4 333 70
GenBank mRNA AJ001261.1 - 3 - 4 311 70
AK170336.1 - 3 - 4 279 70
BC055893.1 - 3 - 4 282 70

View Full Product Details