Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rabgef1
Entrez Gene ID
56715
Gene Name
RAB guanine nucleotide exchange factor (GEF) 1
Gene Aliases
Rab5ef, Rabex5, Rin2
Chromosome Location
Chr.5: 130171791 - 130214342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199059.1 NP_001185988.1 8 - 9 1160 69
NM_019983.2 NP_064367.1 7 - 8 1305 69
GenBank mRNA AF093590.1 - 8 - 9 1136 69
AK044568.1 - 7 - 8 1325 69
AK146734.1 - 8 - 9 1110 69
BC018229.1 - 7 - 8 1305 69

View Full Product Details