Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gatsl2
Entrez Gene ID
80909
Gene Name
GATS protein-like 2
Gene Aliases
7530428J21Rik, AW742413, Gats, Gatsl1
Chromosome Location
Chr.5: 134099748 - 134141782 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030719.3 NP_109644.2 5 - 6 931 76
GenBank mRNA AJ296173.1 - 5 - 6 971 76
AK039122.1 - 5 - 6 957 76
AK052217.1 - 5 - 6 916 76
BC094613.1 - 5 - 6 931 76

View Full Product Details