Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sqle
Entrez Gene ID
20775
Gene Name
squalene epoxidase
Gene Aliases
AI323792
Chromosome Location
Chr.15: 59315092 - 59331193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009270.3 NP_033296.1 9 - 10 2187 103
GenBank mRNA AK138457.1 - 9 - 10 2200 103
AK161837.1 - 8 - 9 2077 103
AK163502.1 - 9 - 10 2167 103
AK208956.1 - 3 - 4 275 103
AK209037.1 - 3 - 4 276 103
AK209538.1 - 3 - 4 275 103
BC042781.1 - 9 - 10 1444 103
BC056361.1 - 9 - 10 2187 103
D42048.1 - 9 - 10 2053 103

View Full Product Details