The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109992.1 | NP_001103462.1 | 10 - 11 | 1408 | 77 |
GenBank mRNA | AK147354.1 | - | 9 - 10 | 3254 | 77 |
AK159501.1 | - | 10 - 11 | 1408 | 77 | |
AK159587.1 | - | 10 - 11 | 1408 | 77 | |
AK163809.1 | - | 7 - 8 | 836 | 77 | |
BC059278.1 | - | 10 - 11 | 1386 | 77 | |
S59187.1 | - | 3 - 4 | 326 | 77 |