The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289543.1 | NP_001276472.1 | 10 - 11 | 1427 | 59 |
NM_001289544.1 | NP_001276473.1 | 11 - 12 | 1751 | 59 | |
NM_028227.3 | NP_082503.2 | 11 - 12 | 1623 | 59 | |
GenBank mRNA | AF321920.1 | - | 11 - 12 | 1515 | 59 |
AF321921.1 | - | 11 - 12 | 1752 | 59 | |
AK013885.1 | - | 9 - 10 | 1116 | 59 | |
AK034212.1 | - | 6 - 7 | 556 | 59 | |
AK143718.1 | - | 10 - 11 | 1417 | 59 | |
BC030384.1 | - | 1 - 2 | 76 | 59 | |
BC038490.1 | - | 8 - 9 | 992 | 59 | |
BC062932.1 | - | 11 - 12 | 1599 | 59 |