The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007756.3 | NP_031782.3 | 3 - 4 | 417 | 60 |
GenBank mRNA | AK134820.1 | - | 3 - 4 | 360 | 60 |
AK158166.1 | - | 3 - 4 | 359 | 60 | |
AK159026.1 | - | 3 - 4 | 376 | 60 | |
BC014803.1 | - | 3 - 4 | 322 | 60 | |
D38614.1 | - | 3 - 4 | 349 | 60 |