Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam49b
Entrez Gene ID
223601
Gene Name
family with sequence similarity 49, member B
Gene Aliases
0910001A06Rik, AW122079
Chromosome Location
Chr.15: 63929094 - 64060452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144846.5 NP_659095.1 10 - 11 1141 69
GenBank mRNA AK028062.1 - 10 - 11 1142 69
AK075613.1 - 11 - 12 1086 69
BC011343.1 - 10 - 11 959 69

View Full Product Details