Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam49b
Entrez Gene ID
223601
Gene Name
family with sequence similarity 49, member B
Gene Aliases
0910001A06Rik, AW122079
Chromosome Location
Chr.15: 63929094 - 64060452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144846.5 NP_659095.1 3 - 4 381 141
GenBank mRNA AK028062.1 - 3 - 4 382 141
AK075613.1 - 4 - 5 326 141
AK205521.1 - 2 - 3 130 141
BC011343.1 - 3 - 4 199 141

View Full Product Details