Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Entpd7
Entrez Gene ID
93685
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
1810012B13Rik, 1810020C02Rik, 2810003F23Rik, LALP1, Lysal2
Chromosome Location
Chr.19: 43689689 - 43733853 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053103.5 NP_444333.3 6 - 7 960 64
GenBank mRNA AK007446.1 - 6 - 7 827 64
AK081564.1 - 6 - 7 818 64
AK147401.1 - 6 - 7 825 64
AK170546.1 - 6 - 7 978 64
BC137709.1 - 5 - 6 746 64
BC137717.1 - 5 - 6 746 64


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF288221.1 - 864 Chr. - - [-] on Build 64

View Full Product Details