Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clcc1
Entrez Gene ID
229725
Gene Name
chloride channel CLIC-like 1
Gene Aliases
Mclc
Chromosome Location
Chr.3: 108653913 - 108678840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177770.1 NP_001171241.1 2 - 3 498 64
NM_001177771.1 NP_001171242.1 3 - 4 413 64
NM_145543.2 NP_663518.1 2 - 3 210 64
GenBank mRNA AK090092.1 - 2 - 3 184 64
AK143631.1 - 3 - 4 396 64
AK169803.1 - 2 - 3 208 64
AK171296.1 - 2 - 3 237 64
BB848444.1 - 2 - 3 211 64
BC003247.1 - 2 - 3 241 64

View Full Product Details