Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psrc1
Entrez Gene ID
56742
Gene Name
proline/serine-rich coiled-coil 1
Gene Aliases
5430413I02Rik, AL033312, Dda3
Chromosome Location
Chr.3: 108383804 - 108388231 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190161.1 NP_001177090.1 3 - 4 199 95
NM_019976.3 NP_064360.1 3 - 4 196 95
GenBank mRNA AF156598.1 - 3 - 4 191 95
AK011197.1 - 3 - 4 171 95
AK017302.1 - 3 - 4 138 95
AK041835.1 - 3 - 4 146 95
AK167415.1 - 3 - 4 142 95
BC019165.1 - 3 - 4 154 95
BY080522.1 - 3 - 4 196 95
BY231608.1 - 3 - 4 199 95

View Full Product Details