Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
50765
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr.5: 137569822 - 137588081 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289509.1 NP_001276438.1 1 - 2 153 62
GenBank mRNA AK004965.1 - 1 - 2 124 62

View Full Product Details