The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256096.1 | NP_001243025.1 | 2 - 3 | 328 | 73 |
NM_001256097.1 | NP_001243026.1 | 3 - 4 | 406 | 73 | |
NM_001256098.1 | NP_001243027.1 | 2 - 3 | 328 | 73 | |
NM_023742.2 | NP_076231.1 | 2 - 3 | 328 | 73 | |
GenBank mRNA | AB015423.1 | - | 2 - 3 | 251 | 73 |
AB015424.1 | - | 2 - 3 | 251 | 73 | |
AK012137.1 | - | 2 - 3 | 324 | 73 | |
AK044105.1 | - | 3 - 4 | 363 | 73 | |
AK054443.1 | - | 2 - 3 | 318 | 73 | |
BC024925.1 | - | 2 - 3 | 270 | 73 | |
CF913708.1 | - | 2 - 3 | 328 | 73 |