Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2rx4
Entrez Gene ID
18438
Gene Name
purinergic receptor P2X, ligand-gated ion channel 4
Gene Aliases
AI504491, AW555605, D5Ertd444e, P2X4
Chromosome Location
Chr.5: 122707504 - 122729739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011026.2 NP_035156.2 6 - 7 697 76
GenBank mRNA AF089751.1 - 6 - 7 631 76
AJ251459.1 - 6 - 7 611 76
AJ251461.1 - 6 - 7 611 76
AK150351.1 - 6 - 7 687 76
AK162820.1 - 6 - 7 663 76
AK163104.1 - 6 - 7 685 76
BC002099.1 - 6 - 7 671 76
JX417451.1 - 6 - 7 657 76
JX674050.1 - 2 - 3 265 76

View Full Product Details