Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lrrc14
- Entrez Gene ID
-
223664
- Gene Name
- leucine rich repeat containing 14
- Gene Aliases
-
E130306I01Rik, mKIAA0014
- Chromosome Location
-
Chr.15: 76710623 - 76715097 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK036278.1 |
- |
1 - 2 |
200 |
75 |
View Full Product Details