The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011223.3 | NP_035353.1 | 5 - 6 | 878 | 77 |
NM_133915.3 | NP_598676.2 | 5 - 6 | 878 | 77 | |
GenBank mRNA | AF293882.1 | - | 5 - 6 | 803 | 77 |
AF293883.1 | - | 4 - 5 | 698 | 77 | |
AK084243.1 | - | 3 - 4 | 543 | 77 | |
AK148060.1 | - | 5 - 6 | 864 | 77 | |
AK149933.1 | - | 5 - 6 | 843 | 77 | |
AK167299.1 | - | 5 - 6 | 817 | 77 | |
AK171226.1 | - | 5 - 6 | 866 | 77 | |
AK171436.1 | - | 5 - 6 | 843 | 77 | |
BC003298.1 | - | 1 - 2 | 161 | 77 | |
BC025493.1 | - | 5 - 6 | 836 | 77 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK089534.1 | - | Chr. - - [-] on Build | 77 |