The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286468.1 | NP_001273397.1 | 7 - 8 | 1892 | 71 |
NM_029270.2 | NP_083546.2 | 9 - 10 | 2494 | 71 | |
NM_146161.3 | NP_666273.1 | 8 - 9 | 2300 | 71 | |
GenBank mRNA | AK080986.1 | - | 9 - 10 | 2494 | 71 |
AK134817.1 | - | 7 - 8 | 2116 | 71 | |
BC023344.1 | - | 7 - 8 | 1906 | 71 | |
BC023440.1 | - | 1 - 2 | 157 | 71 | |
BC025502.1 | - | 8 - 9 | 2149 | 71 | |
BC027070.1 | - | 6 - 7 | 1738 | 71 |