Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scd3
Entrez Gene ID
30049
Gene Name
stearoyl-coenzyme A desaturase 3
Gene Aliases
4930513N16Rik
Chromosome Location
Chr.19: 44203288 - 44244016 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015784.1 - 1 - 2 64 72

View Full Product Details