The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175362.2 | NP_780571.2 | 21 - 22 | 3167 | 59 |
GenBank mRNA | AK037968.1 | - | 7 - 8 | 1032 | 59 |
AK075939.1 | - | 2 - 3 | 347 | 59 | |
AK131134.1 | - | 21 - 22 | 3167 | 59 | |
AY135367.1 | - | 21 - 22 | 3078 | 59 | |
BC096592.1 | - | 21 - 22 | 3153 | 59 |