The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029317.1 | NP_083593.1 | 5 - 6 | 541 | 60 |
GenBank mRNA | AK006268.1 | - | 5 - 6 | 515 | 60 |
AV260738.1 | - | 1 - 2 | 153 | 60 |