Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnf1a
Entrez Gene ID
21405
Gene Name
HNF1 homeobox A
Gene Aliases
AI323641, HNF1, HNF1-alpha, HNF1[a], Hnf-1, Hnf1alpha, LFB1, Tcf-1, Tcf1
Chromosome Location
Chr.5: 114948980 - 114971062 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009327.3 NP_033353.2 9 - 10 1979 106
GenBank mRNA AK141225.1 - 9 - 10 1785 106
AK144051.1 - 9 - 10 1987 106
BC080698.1 - 9 - 10 1981 106
M57966.1 - - 1959 106

View Full Product Details