Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pvt1
- Entrez Gene ID
-
19296
- Gene Name
- plasmacytoma variant translocation 1
- Gene Aliases
-
Gt(pU21)84Imeg, Mis-1, Mlvi-1, Pvt-1
- Chromosome Location
-
Chr.15: 62037987 - 62250975 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090048.1 |
- |
- |
946 |
152 |
View Full Product Details