The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204201.1 | NP_001191130.1 | 1 - 2 | 132 | 64 |
NM_001204202.1 | NP_001191131.1 | 1 - 2 | 132 | 64 | |
GenBank mRNA | AK165965.1 | - | 1 - 2 | 123 | 64 |