Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kank1
Entrez Gene ID
107351
Gene Name
KN motif and ankyrin repeat domains 1
Gene Aliases
A930031B09Rik, AU015049, AW121052, Ankrd15, D330024H06Rik, mKIAA0172
Chromosome Location
Chr.19: 25236732 - 25434498 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181404.5 NP_852069.4 2 - 3 201 80
GenBank mRNA AK020911.1 - 2 - 3 429 80
AK041011.1 - 2 - 3 201 80

View Full Product Details