The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010825.3 | NP_001010825.2 | 2 - 3 | 617 | 80 |
GenBank mRNA | AK044446.1 | - | 2 - 3 | 426 | 80 |
AK054325.1 | - | 2 - 3 | 382 | 80 | |
BC106090.1 | - | 2 - 3 | 588 | 80 | |
BC139821.1 | - | 1 - 2 | 254 | 80 |