The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043354.2 | NP_001036819.1 | 3 - 4 | 871 | 66 |
NM_001289921.1 | NP_001276850.1 | 4 - 5 | 390 | 66 | |
NM_146095.4 | NP_666207.3 | 3 - 4 | 337 | 66 | |
GenBank mRNA | AB258405.1 | - | 3 - 4 | 498 | 66 |
AK044421.1 | - | 3 - 4 | 332 | 66 | |
AK134790.1 | - | 3 - 4 | 637 | 66 | |
BC024768.1 | - | 4 - 5 | 373 | 66 | |
BC024842.1 | - | 4 - 5 | 390 | 66 | |
BC058269.1 | - | 2 - 3 | 244 | 66 | |
DQ779923.1 | - | 2 - 3 | 241 | 66 | |
DQ779924.1 | - | 3 - 4 | 266 | 66 |