Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tnks
- Entrez Gene ID
-
21951
- Gene Name
- tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase
- Gene Aliases
-
4930554K12Rik, AI662855, ARTD5, C86528, D130072O21Rik, TANK1, mTNKS1
- Chromosome Location
-
Chr.8: 34829179 - 34965690 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175091.3 |
NP_780300.2 |
1 - 2 |
676 |
78 |
GenBank mRNA |
BC057370.1 |
- |
1 - 2 |
657 |
78 |
View Full Product Details