The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033229.3 | NP_001028401.2 | 5 - 6 | 994 | 81 |
GenBank mRNA | AK142439.1 | - | 5 - 6 | 971 | 81 |
AK163083.1 | - | 5 - 6 | 970 | 81 | |
AK168839.1 | - | 5 - 6 | 971 | 81 | |
BC139248.1 | - | 5 - 6 | 994 | 81 | |
BC139250.1 | - | 5 - 6 | 994 | 81 |