The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033174.1 | NP_001028346.1 | 9 - 10 | 1765 | 74 |
GenBank mRNA | AK132663.1 | - | 7 - 8 | 1045 | 74 |
AK220301.1 | - | 6 - 7 | 803 | 74 | |
BC094342.1 | - | 3 - 4 | 386 | 74 | |
BC107327.1 | - | 9 - 10 | 1499 | 74 | |
BC107328.2 | - | 9 - 10 | 1499 | 74 |