Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpsf1
Entrez Gene ID
94230
Gene Name
cleavage and polyadenylation specific factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 76595808 - 76607613 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164173.1 NP_001157645.1 17 - 18 1864 94
NM_053193.2 NP_444423.1 17 - 18 1864 94
GenBank mRNA AF322193.1 - 17 - 18 1835 94
AK149914.1 - 10 - 11 863 94
AK155648.1 - 17 - 18 1805 94
AK155806.1 - 17 - 18 1786 94
BC011458.1 - 13 - 14 1303 94
BC049941.1 - 17 - 18 1686 94
BC056388.1 - 17 - 18 1864 94
BC057945.1 - 2 - 3 195 94

View Full Product Details