The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172723.4 | NP_766311.2 | 1 - 2 | 188 | 111 |
GenBank mRNA | BY740716.1 | - | 1 - 2 | 346 | 111 |
CX242691.1 | - | 1 - 2 | 188 | 111 |