The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163316.1 | NP_001156788.1 | 5 - 6 | 554 | 79 |
NM_026269.2 | NP_080545.2 | 5 - 6 | 745 | 79 | |
GenBank mRNA | AK003551.1 | - | 5 - 6 | 694 | 79 |
BC019557.1 | - | 5 - 6 | 536 | 79 |