The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111058.1 | NP_001104528.1 | 4 - 5 | 865 | 77 |
NM_021293.3 | NP_067268.1 | 4 - 5 | 865 | 77 | |
GenBank mRNA | AK046303.1 | - | 6 - 7 | 1088 | 77 |
AK155745.1 | - | 4 - 5 | 795 | 77 | |
AK170937.1 | - | 4 - 5 | 833 | 77 | |
BC132379.1 | - | 4 - 5 | 865 | 77 | |
S71345.1 | - | 4 - 5 | 796 | 77 | |
S71403.1 | - | 4 - 5 | 796 | 77 |