The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010246.1 | NP_034376.1 | 1 - 1 | 2065 | 68 |
GenBank mRNA | AF033585.2 | - | 1 - 1 | 1920 | 68 |
AK021164.1 | - | 1 - 1 | 2249 | 68 |