Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnah10
Entrez Gene ID
56087
Gene Name
dynein, axonemal, heavy chain 10
Gene Aliases
Dnahc10
Chromosome Location
Chr.5: 124725018 - 124834827 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019536.1 NP_062409.1 25 - 26 4475 71

View Full Product Details