Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp3a13
Entrez Gene ID
13113
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 13
Gene Aliases
IIIAm2
Chromosome Location
Chr.5: 137892933 - 137921619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007819.4 NP_031845.1 12 - 13 1519 72
GenBank mRNA AK136494.1 - 12 - 13 1518 72
AK136529.1 - 12 - 13 1520 72
AK149572.1 - 12 - 13 1520 72
BC046592.1 - 12 - 13 1498 72
X63023.1 - 12 - 13 1495 72

View Full Product Details