Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
11593
Gene Name
aspartylglucosaminidase
Gene Aliases
AW060726
Chromosome Location
Chr.8: 53511687 - 53523422 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005847.2 NP_001005847.1 2 - 3 382 61
NM_001205054.1 NP_001191983.1 2 - 3 382 61
GenBank mRNA AA396103.1 - 2 - 3 60
BC048230.1 - 2 - 3 1263 61
BC132227.1 - 2 - 3 357 61
BC132229.1 - 2 - 3 357 61
BC145297.1 - 2 - 3 357 61
BQ571087.1 - 2 - 3 274 61
CD541877.1 - 2 - 3 358 61
S81393.1 - 2 - 3 299 61

View Full Product Details