The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027891.4 | NP_082167.2 | 4 - 5 | 700 | 79 |
GenBank mRNA | AK004717.1 | - | 3 - 4 | 1116 | 79 |
AK051910.1 | - | 4 - 5 | 648 | 79 | |
AK084948.1 | - | 4 - 5 | 700 | 79 | |
AK153976.1 | - | 4 - 5 | 671 | 79 | |
AK154352.1 | - | 4 - 5 | 642 | 79 | |
AK167020.1 | - | 4 - 5 | 649 | 79 | |
BC048397.1 | - | 4 - 5 | 679 | 79 |