The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183216.3 | NP_899039.2 | 2 - 3 | 452 | 61 |
GenBank mRNA | AY430080.1 | - | 2 - 3 | 452 | 61 |
BC141234.1 | - | 2 - 3 | 408 | 61 |