Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aptx
Entrez Gene ID
66408
Gene Name
aprataxin
Gene Aliases
2410016G21Rik, AA388047, FHA-HIT
Chromosome Location
Chr.4: 40682078 - 40721667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025445.2 NP_001020616.1 2 - 3 198 92
GenBank mRNA AK010516.1 - 2 - 3 185 92
AK017522.1 - 1 - 2 168 92
BC068309.1 - 1 - 2 111 92

View Full Product Details