Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2d4b
Entrez Gene ID
328381
Gene Name
SH2 domain containing 4B
Gene Aliases
A430109M18Rik, D030001E08
Chromosome Location
Chr.14: 40811521 - 40902337 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177816.3 NP_808484.2 3 - 4 493 70
GenBank mRNA AK020787.1 - 3 - 4 799 70
AX776199.1 - 3 - 4 493 70
BC138964.1 - 3 - 4 591 70

View Full Product Details