Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem68
Entrez Gene ID
72098
Gene Name
transmembrane protein 68
Gene Aliases
2010300G19Rik, AA408325, AW123402
Chromosome Location
Chr.4: 3549041 - 3574848 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028097.3 NP_082373.1 6 - 7 922 120
GenBank mRNA AK008493.1 - 6 - 7 956 120
AK048210.1 - 6 - 7 922 120
AK084881.1 - 6 - 7 956 120
AK088600.1 - 7 - 8 1092 120
AK147765.1 - 6 - 7 955 120
AK162563.1 - 6 - 7 955 120
AK165916.1 - 6 - 7 984 120
AK166237.1 - 6 - 7 1008 120
BC016240.1 - 6 - 7 940 120

View Full Product Details