The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011292.2 | NP_035422.1 | 2 - 3 | 106 | 60 |
GenBank mRNA | AF260271.1 | - | 1 - 2 | 76 | 60 |
AK012331.1 | - | 2 - 3 | 76 | 60 | |
AK012445.1 | - | 2 - 3 | 76 | 60 | |
AK017388.1 | - | 2 - 3 | 76 | 60 | |
AK017427.1 | - | 2 - 3 | 76 | 60 | |
AK053431.1 | - | 1 - 2 | 459 | 60 | |
AK084278.1 | - | 2 - 3 | 77 | 60 | |
AK088166.1 | - | 2 - 3 | 76 | 60 | |
AK153562.1 | - | 2 - 3 | 76 | 60 | |
AK159980.1 | - | 2 - 3 | 76 | 60 | |
AK168326.1 | - | 2 - 3 | 74 | 60 | |
AK169217.1 | - | 2 - 3 | 74 | 60 | |
BC013165.1 | - | 1 - 2 | 66 | 60 | |
BC081435.1 | - | 1 - 2 | 52 | 60 | |
BC083166.1 | - | 1 - 2 | 57 | 60 | |
BC083329.1 | - | 1 - 2 | 55 | 60 | |
BC089319.1 | - | 1 - 2 | 55 | 60 | |
BC093526.1 | - | 1 - 2 | 55 | 60 |