Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kirrel
Entrez Gene ID
170643
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
6720469N11Rik, Kirrel1, Neph1
Chromosome Location
Chr.3: 87078592 - 87174747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170985.1 NP_001164456.1 2 - 3 498 78
NM_130867.3 NP_570937.2 2 - 3 495 78
GenBank mRNA AF480411.1 - 1 - 2 133 78
AK158381.1 - 2 - 3 452 78
AY017368.1 - 1 - 2 133 78
AY243095.1 - 1 - 2 133 78
BC023765.1 - 2 - 3 311 78
BY736743.1 - 2 - 3 281 78

View Full Product Details