Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sftpa1
Entrez Gene ID
20387
Gene Name
surfactant associated protein A1
Gene Aliases
SP-A, Sftp-1, Sftp1
Chromosome Location
Chr.14: 41131787 - 41136373 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023134.4 NP_075623.2 5 - 6 476 76
GenBank mRNA AK004620.1 - 5 - 6 477 76
AK004788.1 - 5 - 6 477 76
AK011333.1 - 4 - 5 450 76
AK075702.1 - 5 - 6 480 76
BC061096.1 - 5 - 6 293 76

View Full Product Details